Acetylcholine iodide 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(2-Acetoxyethyl)trimethylammonium iodide
A11694-14
A11694-14
AAA11694-14EA
358.99
USD
AAA11694-14
CAAAA11694-14
Acetylcholine iodide 98%
Acetylcholine iodide
Formula:
H₃CC(O)O(CH₂)₂N(CH₃)₃I MW: 273.11 g/mol Melting Pt: 161…165 °C Density: 1,67 g/cm³ (20 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00011815 CAS Number: 2260-50-6 EINECS: 218-862-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...