2,6-Difluorobenzamide 98+%
|
Warning
|
A11685-22
A11685-14
A11685-22
A11685-14
AAA11685-14EA
59.5
USD
CAAAA11685-22
AAA11685-14
CAAAA11685-14
AAA11685-22
2,6-Difluorobenzamide 98+%
2,6-Difluorobenzamide
Formula:
C₇H₅F₂NO Boiling Pt: 284 °C (1013 hPa) Melting Pt: 145…148 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007972 CAS Number: 18063-03-1 EINECS: 241-972-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...