3-Nitro-p-toluic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Methyl-3-nitrobenzoic acid

A11650-36 A11650-22 A11650-22 A11650-36
AAA11650-22EA 53.19 USD
CAAAA11650-22 AAA11650-22 AAA11650-36 CAAAA11650-36
3-Nitro-p-toluic acid 99%
3-Nitro-p-toluic acid
Formula: C₈H₇NO₄
MW: 181.15 g/mol
Boiling Pt: 360 °C (1013 hPa)
Melting Pt: 188…190 °C
Storage Temperature: Ambient
MDL Number: MFCD00007174
CAS Number: 96-98-0
EINECS: 202-549-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR