2,3-Dimethylbenzonitrile 97%

Warning

A11589-14 A11589-14 A11589-06 A11589-06
AAAA11589-14EA 336.33 USD
CAAAAA11589-14 AAAA11589-14 CAAAA11589-06 AAA11589-06
2,3-Dimethylbenzonitrile 97%
2,3-Dimethylbenzonitrile
Formula: C₉H₉N
MW: 131.18 g/mol
Boiling Pt: 64…66 °C/0.8 mm
Melting Pt: 23…25 °C
Storage Temperature: Ambient
MDL Number: MFCD00019744
CAS Number: 5724-56-1
EINECS: 227-229-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR