p-Toluic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Methylbenzoic acid 4-Toluic acid

A11506-0B A11506-0I A11506-30 A11506-0I A11506-30 A11506-0B
AAA11506-30EA 52.05 USD
AAA11506-30 CAAAA11506-0B CAAAA11506-30 AAA11506-0B CAAAA11506-0I AAA11506-0I
p-Toluic acid 98%
p-Toluic acid
Formula: C₈H₈O₂
MW: 136.15 g/mol
Boiling Pt: 274 °C
Melting Pt: 178…182 °C
Density: 1,06
Flash Pt: 181 °C(357 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002565
CAS Number: 99-94-5
EINECS: 202-803-3
Merck Index: 12,09673

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR