1-Hexadecyne tech. 90%, Technical Grade

Supplier: Thermo Scientific Chemicals

A11366-14 A11366-06 A11366-14 A11366-06
AAA11366-14EA 444.99 USD
CAAAA11366-14 CAAAA11366-06 AAA11366-14 AAA11366-06
1-Hexadecyne tech. 90%, Technical Grade
1-Hexadecyne
Formula: C₁₆H₃₀
MW: 222.41 g/mol
Boiling Pt: 146…148 °C/10 mm
Melting Pt: 13…16 °C
Density: 0.790
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00015084
CAS Number: 629-74-3
EINECS: 211-106-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR