4-Amino-3,5-dichlorobenzonitrile 97%

Synonyms: 4-Cyano-2,6-dichloroaniline

A11269-06 A11269-14 A11269-14 A11269-03 A11269-06 A11269-03
AAA11269-14EA 833.45 USD
CAAAA11269-14 CAAAA11269-03 AAA11269-14 AAA11269-03 AAA11269-06 CAAAA11269-06
4-Amino-3,5-dichlorobenzonitrile 97%
4-Amino-3,5-dichlorobenzonitrile
Formula: C₇H₄Cl₂N₂
MW: 187.03 g/mol
Melting Pt: 117…119 °C
Storage Temperature: Ambient
MDL Number: MFCD00017341
CAS Number: 78473-00-4
EINECS: 278-914-6
UN: 3439
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR