4-Chloro-4'-fluorobutyrophenone 96%

Warning

Synonyms: γ-Chloro-4-fluorobutyrophenone ω-Chloro-4-fluorobutyrophenone

A11121-14 A11121-36 A11121-36 A11121-14 A11121-22 A11121-22
AAA11121-36EA 292.57 USD
AAA11121-36 AAA11121-14 AAA11121-22 CAAAA11121-22 CAAAA11121-14 CAAAA11121-36
4-Chloro-4'-fluorobutyrophenone 96%
4-Chloro-4'-fluorobutyrophenone
Formula: C₁₀H₁₀ClFO
MW: 200.64 g/mol
Boiling Pt: 120…122 °C/0.7 mm
Melting Pt: 5…6 °C
Density: 1.23
Flash Pt: 152 °C(306 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00001007
CAS Number: 3874-54-2
EINECS: 223-395-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR