α,3-Dichlorotoluene 97%
|
Danger
|
Synonyms:
1-Chloro-3-(chloromethyl)benzene
A11041-14
A11041-22
A11041-22
A11041-14
AAA11041-14EA
47.18
USD
CAAAA11041-14
CAAAA11041-22
AAA11041-14
AAA11041-22
α,3-Dichlorotoluene 97%
α,3-Dichlorotoluene
Formula:
C₇H₆Cl₂ MW: 161.03 g/mol Boiling Pt: 215…216 °C Melting Pt: 69…71 °C Density: 1.270 Flash Pt: 98 °C(208 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000905 CAS Number: 620-20-2 EINECS: 210-629-4 UN: 2235 ADR: "6.1",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...