Tetrahydrothiophen-3-one 97%

Warning

Synonyms: 4,5-Dihydrothiophen-3(2H)-one

A11002-09 A11002-30 A11002-30 A11002-06 A11002-06 A11002-14 A11002-14 A11002-09
AAAA11002-06EA 30.08 USD
AAAA11002-06 AAA11002-09 CAAAA11002-30 CAAAA11002-09 CAAAAA11002-06 CAAAAA11002-14 AAA11002-30 AAAA11002-14
Tetrahydrothiophen-3-one 97%
Tetrahydrothiophen-3-one
Formula: C₄H₆OS
MW: 102.16 g/mol
Boiling Pt: 174…175 °C
Density: 1,192
Flash Pt: 77 °C(170 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005412
CAS Number: 1003-04-9
EINECS: 213-698-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR