Dicyclohexylcarbodiimide 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
N,N'-Dicyclohexylcarbodiimide
A10973-14
A10973-36
A10973-22
A10973-14
A10973-22
A10973-36
AAA10973-14EA
51.27
USD
AAA10973-14
AAA10973-36
AAA10973-22
CAAAA10973-14
CAAAA10973-36
CAAAA10973-22
Dicyclohexylcarbodiimide 99%
Dicyclohexylcarbodiimide
Carboxy group activating reagent for peptide synthesis
Formula:
C₁₃H₂₂N₂ MW: 206.33 g/mol Boiling Pt: 154…156 °C/11 mm Melting Pt: 33…36 °C Flash Pt: 138 °C(280 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00011659 CAS Number: 538-75-0 EINECS: 208-704-1 UN: 2811 ADR: "6.1",III Merck Index: 13,03123 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...