1-Phenyl-2-hexanone 95%

Supplier: Thermo Scientific Chemicals

A10944-06 A10944-14 A10944-03 A10944-14 A10944-03 A10944-06
AAA10944-03EA 32.89 USD
AAA10944-03 CAAAA10944-03 CAAAAA10944-14 AAAA10944-14 CAAAA10944-06 AAA10944-06
1-Phenyl-2-hexanone 95%
1-Phenyl-2-hexanone
Formula: C₆H₅CH₂C(O)(CH₂)₃CH₃
MW: 176.26 g/mol
Boiling Pt: 153 °C/20 mm
Density: 0.940
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00027240
CAS Number: 25870-62-6
EINECS: 247-306-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR