3-Fluoroanisole 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Fluoro-3-methoxybenzene

A10931-09 A10931-09 A10931-30 A10931-30
AAA10931-30EA 422.62 USD
AAA10931-30 AAA10931-09 CAAAA10931-09 CAAAA10931-30
3-Fluoroanisole 99%
3-Fluoroanisole
Formula: C₇H₇FO
MW: 126.13 g/mol
Boiling Pt: 158…159 °C
Melting Pt: –35 °C
Density: 1.104
Flash Pt: 43 °C(109 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000335
CAS Number: 456-49-5
EINECS: 207-267-4
UN: 1993
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR