3-Aminobenzamide 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: m-AminobenzamideINO-1001

A10793-06 A10793-06 A10793-14 A10793-14
AAA10793-06EA 64.19 USD
CAAAA10793-06 AAA10793-06 CAAAA10793-14 AAA10793-14
3-Aminobenzamide 98%
3-Aminobenzamide

Endogenous poly-ADP-ribosyltransferase inhibitor

Formula: C₇H₈N₂O
MW: 136.15 g/mol
Melting Pt: 111…115 °C
Storage Temperature: Ambient
MDL Number: MFCD00007989
CAS Number: 3544-24-9
EINECS: 222-586-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR