m-Cresol 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1-Hydroxy-3-methylbenzene 3-Hydroxytoluene 3-Methylphenol

A10733-30 A10733-30 A10733-0B A10733-0E A10733-0B A10733-0E
AAA10733-0BEA 115.22 USD
AAA10733-0B AAA10733-0E CAAAA10733-30 CAAAA10733-0B AAA10733-30 CAAAA10733-0E
m-Cresol 99%
m-Cresol
Formula: CH₃C₆H₄OH
MW: 108.14 g/mol
Boiling Pt: 202…204 °C
Melting Pt: 11…12 °C
Density: 1.034
Flash Pt: 86 °C(186 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002302
CAS Number: 108-39-4
EINECS: 203-577-9
UN: 2076
ADR: "6.1",II
Merck Index: 13,02604
https://us.vwr.com/stibo/web/std.lang.all/08/21/2300821.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR