N,N,2,4,6-Pentamethylaniline 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2,4,6,N,N-Pentamethylaniline

A10561-22 A10561-14 A10561-06 A10561-14 A10561-36 A10561-06 A10561-36 A10561-22
AAA10561-22EA 273.72 USD
CAAAA10561-14 CAAAA10561-36 AAA10561-22 CAAAA10561-06 AAA10561-14 AAA10561-36 CAAAA10561-22 AAA10561-06
N,N,2,4,6-Pentamethylaniline 98%
N,N,2,4,6-Pentamethylaniline
Formula: C₁₁H₁₇N
MW: 163.26 g/mol
Boiling Pt: 213…215 °C
Density: 0,907
Flash Pt: 78 °C(174 °F)
Storage Temperature: Ambient
MDL Number: MFCD00010596
CAS Number: 13021-15-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR