2-Chloro-4-fluorobenzenesulfonyl chloride 98%

Danger

Synonyms: 2-Chloro-4-fluorobenzene-1-sulphonyl chloride

A10536-14 A10536-03 A10536-03 A10536-06 A10536-14 A10536-06
AAAA10536-14EA 520.59 USD
CAAAA10536-03 CAAAAA10536-14 CAAAA10536-06 AAAA10536-14 AAA10536-03 AAA10536-06
2-Chloro-4-fluorobenzenesulfonyl chloride 98%
2-Chloro-4-fluorobenzenesulphonyl chloride
Formula: C₆H₃Cl₂FO₂S
MW: 229.06 g/mol
Boiling Pt: 243…244 °C
Melting Pt: 28…31 °C
Density: 1.57
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00052957
CAS Number: 85958-57-2
UN: 3265
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR