2-Chloro-6-fluorobenzaldehyde 97%

Supplier: Thermo Scientific Chemicals
Warning

A10513-22 A10513-14 A10513-14 A10513-22
AAA10513-22EA 96.69 USD
AAA10513-22 CAAAA10513-22 CAAAA10513-14 AAA10513-14
2-Chloro-6-fluorobenzaldehyde 97%
2-Chloro-6-fluorobenzaldehyde
Formula: C₇H₄ClFO
MW: 158.56 g/mol
Boiling Pt: 91…93 °C/10 mm
Melting Pt: 35…39 °C
Flash Pt: 101 °C(213 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003306
CAS Number: 387-45-1
EINECS: 206-860-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR