Bis(2-ethylhexyl)phthalate 98+%

Danger

Synonyms: Phthalic acid bis(2-ethylhexyl) ester DEHP Di-iso-octyl phthalate Di-(2-ethylhexyl) phthalate Di(2-ethylhexyl) phthalate

A10415-36 A10415-0C A10415-0E A10415-0C A10415-0E
AAA10415-0EEA 99.18 USD
CAAAA10415-36 CAAAA10415-0C CAAAA10415-0E AAA10415-0E AAA10415-0C
Bis(2-ethylhexyl)phthalate 98+%
Bis(2-ethylhexyl) phthalate
Formula: C₂₄H₃₈O₄
MW: 390.56 g/mol
Boiling Pt: 230 °C/5 mm
Melting Pt: –55 °C
Density: 0.985
Flash Pt: 207 °C(404 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009493
CAS Number: 117-81-7
EINECS: 204-211-0
ADR: 9,III
Merck Index: 13,02884
https://us.vwr.com/stibo/web/std.lang.all/70/80/2297080.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR