Hexamethylbenzene 99+%

Supplier: Thermo Scientific Chemicals

A10294-03 A10294-06 A10294-14 A10294-06 A10294-03 A10294-14
AAAA10294-14EA 297.21 USD
CAAAAA10294-14 AAAA10294-14 CAAAA10294-06 AAA10294-03 CAAAA10294-03 AAA10294-06
Hexamethylbenzene 99+%
Hexamethylbenzene
Formula: C₁₂H₁₈
MW: 162.27 g/mol
Boiling Pt: 265 °C
Melting Pt: 163…167 °C
Density: 1.063
Storage Temperature: Ambient
MDL Number: MFCD00008523
CAS Number: 87-85-4
EINECS: 201-777-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR