2,3,4-Trifluoroaniline 98%

Danger

A10254-14 A10254-14 A10254-22 A10254-22
AAA10254-22EA 420.11 USD
CAAAA10254-14 CAAAA10254-22 AAA10254-22 AAA10254-14
2,3,4-Trifluoroaniline 98%
2,3,4-Trifluoroaniline
Formula: C₆H₄F₃N
MW: 147.1 g/mol
Boiling Pt: 90…92 °C/48 mm
Melting Pt: 14…15 °C
Density: 1.393
Flash Pt: 68 °C(154 °F)
Storage Temperature: Ambient
MDL Number: MFCD00011737
CAS Number: 3862-73-5
EINECS: 407-170-9
UN: 2941
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR