3,5-Dichloro-2,4,6-trifluoropyridine 97+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3,5-Dichlorotrifluoropyridine

A10205-14 A10205-06 A10205-22
AAA10205-14EA 118.99 USD
AAA10205-14 AAAA10205-22 AAA10205-06
3,5-Dichloro-2,4,6-trifluoropyridine 97+%
3,5-Dichloro-2,4,6-trifluoropyridine
Formula: C₅Cl₂F₃N
MW: 201.96 g/mol
Boiling Pt: 159…160 °C
Melting Pt: 24…26 °C
Density: 1.648
Flash Pt: None
Storage Temperature: Ambient
MDL Number: MFCD00006226
CAS Number: 1737-93-5
EINECS: 217-088-3
UN: 3382
ADR: "6.1",I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR