Sodium hydroxide concentrate for dilution to 0.1 N in aqueous solution, Acculute™ Standard Volumetric Solution
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Sodium hydrate
, Soda lye
, Caustic soda
, Lye
088623-LQ
88623-LQ
AA88623-LQEA
67.09
USD
AA88623-LQ
CAAA88623-LQ
Sodium hydroxide concentrate for dilution to 0.1 N in aqueous solution, Acculute™ Standard Volumetric Solution
Sodium hydroxide
Formula:
NaOH MW: 40 g/mol Boiling Pt: ∼105 °C (1013 hPa) Density: 1.327 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003548 CAS Number: 1310-73-2 EINECS: 215-185-5 UN: 1824 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...