Acetic acid concentrate for dilution to 1.0 N in aqueous solution, Acculute™ standard volumetric solution

Synonyms: Methanecarboxylic acidVinegar acidEthanoic acid

88556-LS 88556-LS
AA88556-LSEA 405.51 USD
CAAA88556-LS AA88556-LS
Acetic acid concentrate for dilution to 1.0 N in aqueous solution, Acculute™ standard volumetric solution
Acetic acid
Formula: H₃CCOOH
MW: 60.05 g/mol
MDL Number: MFCD00036152
CAS Number: 64-19-7
UN: 2790
ADR: 8,III
https://us.vwr.com/stibo/web/std.lang.all/31/15/723115.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR