Sodium hexafluoroarsenate 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Sodium hexafluoroarsenate(V)
88517-09
088517-09
AA88517-09EA
245.99
USD
CAAA88517-09
AA88517-09
Sodium hexafluoroarsenate 99%
Sodium hexafluoroarsenate
Formula:
AsF₆Na MW: 211.9 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00014228 CAS Number: 12005-86-6 UN: 1557 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...