Sodium hexafluoroaluminate ≥97.8%

Supplier: Thermo Scientific Chemicals

Danger

Synonyms: Trisodium hexafluoroaluminate

88275-36 88275-36
AA88275-36EA 102 USD
AA88275-36 CAAA88275-36
Sodium hexafluoroaluminate ≥97.8%
Sodium hexafluoroaluminate
Formula: AlF₆Na₃
MW: 209.94 g/mol
Melting Pt: 1000 °C
Density: 2.90
Storage Temperature: Ambient
MDL Number: MFCD00003507
CAS Number: 13775-53-6
EINECS: 237-410-6
UN: 3077
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR