Thallium(I) ethanolate 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Thallium (I) ethoxide

78131-06 078131-06
AA78131-06EA 151.99 USD
AA78131-06 CAAA78131-06
Thallium(I) ethanolate 98%
Thallium (I) ethanolate
Formula: C₂H₅OTl
MW: 249.44 g/mol
Boiling Pt: 130 °C (dec.)
Melting Pt: –3 °C
Density: 3,493
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009072
CAS Number: 20398-06-5
EINECS: 243-786-2
UN: 2810
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR