Tetraphenyltin

Danger

Synonyms: Tetraphenylstannane

71117-22 71117-22
AA71117-22EA 71.89 USD
AA71117-22 CAAA71117-22
Tetraphenyltin
Tetraphenyltin
Formula: C₂₄H₂₀Sn
MW: 427.13 g/mol
Boiling Pt: >420 °C
Melting Pt: 224…227 °C
Density: 1,49 g/cm³ (5 °C)
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003006
CAS Number: 595-90-4
EINECS: 209-872-9
UN: 3146
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR