Decacarbonyldirhenium ≥96%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Rhenium carbonyl

62108-03 062108-03
AA62108-03EA 137.99 USD
CAAA62108-03 AA62108-03
Decacarbonyldirhenium ≥96%
Decacarbonyldirhenium
Formula: C₁₀O₁₀Re₂
MW: 652.52 g/mol
Melting Pt: 170 °C (dec.)
Density: 2,87
Storage Temperature: Ambient
MDL Number: MFCD00011198
CAS Number: 14285-68-8
EINECS: 238-202-8
UN: 3466
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR