Strontium titanate substrate 5x5x0.5 mm, polished both sides 100 orientation

Synonyms: Strontium titanate Strontium titanate (IV) Strontium titanium oxide

42663-AE 42668-KU 42668-KT 42668-KU 42668-KT
AA42668-KUEA 492.64 USD
CAAA42668-KU CAAA42668-KT CAAA42663-AE AA42668-KU AA42668-KT
Strontium titanate substrate 5x5x0.5 mm, polished both sides 100 orientation
Strontium titanate
Formula: O₃SrTi
Melting Pt: 2080 °C
Density: 5.11 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00049554
CAS Number: 12060-59-2
EINECS: 235-044-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR