Strontium titanate substrate 6.25x6.25x0.5 mm, polished one side 110 orientation

Synonyms: Strontium titanate Strontium titanate (IV) Strontium titanium oxide

42617-KT 42617-KT 42617-KU 42617-KU
AA42617-KTEA 133.2 USD
AA42617-KT AA42617-KU CAAA42617-KT CAAA42617-KU
Strontium titanate substrate 6.25x6.25x0.5 mm, polished one side 110 orientation
Strontium titanate
Formula: O₃SrTi
Melting Pt: 2080 °C
Density: 5.11 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00049554
CAS Number: 12060-59-2
EINECS: 235-044-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR