2,2,2-Trifluoroethanol-[D₃] (100% D) (Isotopic) for NMR spectroscopy

Danger

Synonyms: TFE-D3 2,2,2-Trifluoroethanol-[²H₃]

42364-03 42364-03 42364-06 42364-06
AA42364-06EA 814.74 USD
AA42364-06 CAAA42364-03 CAAA42364-06 AA42364-03
2,2,2-Trifluoroethanol-[D₃] (100% D) (Isotopic) for NMR spectroscopy
2,2,2-Trifluoroethanol-[D3]

NMR Solvent

Formula: F₃CCD₂OD
MW: 103.02 g/mol
Boiling Pt: 77…80 °C
Melting Pt: –44 °C
Density: 1.45
Storage Temperature: Ambient
MDL Number: MFCD00037682
CAS Number: 77253-67-9
EINECS: 278-649-6
UN: 1992
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR