Nitromethane-[D₃] (99% D) (Isotopic) for NMR spectroscopy

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Nitromethane-[²H₃]

42338.06 42338-06
AA42338-06EA 94.44 USD
AA42338-06 CAAA42338-06
Nitromethane-[D₃] (99% D) (Isotopic) for NMR spectroscopy
Nitromethane-[D3]
Formula: D₃CNO₂
MW: 64.02 g/mol
Boiling Pt: 100 °C
Melting Pt: –26 °C
Density: 1.183
Flash Pt: 44 °C
Storage Temperature: Ambient
MDL Number: MFCD00044214
CAS Number: 13031-32-8
EINECS: 235-892-2
UN: 1261
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR