Bromopentacarbonylrhenium 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Rhenium pentacarbonyl bromide
, Bromopentacarbonylrhenium(I)
42097-03
042097-03
AA42097-03EA
323.99
USD
AA42097-03
CAAA42097-03
Bromopentacarbonylrhenium 98%
Bromopentacarbonylrhenium
Formula:
C₅BrO₅Re MW: 406.16 g/mol Melting Pt: ∼ 90 °C (subl.) Storage Temperature: Ambient |
MDL Number:
MFCD00013295 CAS Number: 14220-21-4 EINECS: 238-084-8 UN: 3466 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...