Aluminium magnesium silicon copper chromium alloy (97.93:1.0:0.6:0.27:0.2 w%), alloy 6061, rods 3.175 mm (0.125 in) Ø, Diameter 3.175 mm (0.125 in)
Synonyms: Alloy 6061
42051-CE 42051-BV 42051-BV 42051-CE
AA42051-CEEA 35.2 USD
AA42051-CE CAAA42051-BV AA42051-BV CAAA42051-CE
Aluminium magnesium silicon copper chromium alloy (97.93:1.0:0.6:0.27:0.2 w%), alloy 6061, rods 3.175 mm (0.125 in) Ø, Diameter 3.175 mm (0.125 in)
Aluminium Magnesium Silicon Copper Chromium alloy (97.93:1.0:0.6:0.27:0.2 w%), alloy 6061