2-Hydroxyacetophenone 97+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Phenacyl hydroxyide α-Hydroxyacetophenone 2-Hydroxyacetophenone

42013-06 042013-06
AA42013-06EA 81.69 USD
CAAA42013-06 AA42013-06
2-Hydroxyacetophenone 97+%
2-Hydroxyacetophenone
Formula: C₈H₈O₂
MW: 136.15 g/mol
Boiling Pt: 118…120 °C (11 mmHg)
Melting Pt: 86…88 °C
Density: 1.0963 g/cm³ (99 °C)
Storage Temperature: Ambient
MDL Number: MFCD00041829
CAS Number: 582-24-1
EINECS: 209-480-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR