Silicon(II) oxide ≥99.9% (metals basis)

Supplier: Thermo Scientific Chemicals

Synonyms: Silicium (II) oxide Silicon monoxide Silicon oxide

041711-22 41711-22
AA41711-22EA 201.99 USD
CAAA41711-22 AA41711-22
Silicon(II) oxide ≥99.9% (metals basis)
Silicon (II) oxide

For vacuum deposition

Formula: SiO
MW: 44.08 g/mol
Boiling Pt: 1880 °C
Melting Pt: >1702 °C
Density: 2.13
Storage Temperature: Ambient
MDL Number: MFCD00151536
CAS Number: 10097-28-6
EINECS: 233-232-8
Merck Index: 12,08640

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR