Arsenic(III) telluride ≥99.999% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

041708-06 41708-06
AA41708-06EA 129.99 USD
CAAA41708-06 AA41708-06
Arsenic(III) telluride ≥99.999% (metals basis)
Arsenic (III) telluride
Formula: As₂Te₃
MW: 532.64 g/mol
Melting Pt: 381 °C
Density: 6.50
Storage Temperature: Ambient
MDL Number: MFCD00014167
CAS Number: 12044-54-1
EINECS: 234-955-1
UN: 1557
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR