Praseodymium trifluoride, anhydrous ≥99.9% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Praseodymium (III) fluoride

41418-18 41418-09 041418-18 41418-09
AA41418-09EA 98.4 USD
AA41418-09 CAAA41418-18 CAAA41418-09 AA41418-18
Praseodymium trifluoride, anhydrous ≥99.9% (metals basis)
Praseodymium trifluoride

For vacuum deposition

Formula: F₃Pr
MW: 197.9 g/mol
Melting Pt: 1395 °C
Density: 6.2
Storage Temperature: Ambient
MDL Number: MFCD00016287
CAS Number: 13709-46-1
EINECS: 237-254-9
UN: 3288
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR