Lutetium trifluoride, anhydrous ≥99.9% (REO, rare earth oxide basis)

Danger

Synonyms: Lutetium (III) fluoride

41415-03 41415-03
AA41415-03EA 218.67 USD
CAAA41415-03 AA41415-03
Lutetium trifluoride, anhydrous ≥99.9% (REO, rare earth oxide basis)
Lutetium trifluoride

For vacuum deposition

Formula: LuF₃
MW: 231.96 g/mol
Boiling Pt: 2200 °C (1013 hPa)
Melting Pt: 1182 °C
Density: 8.3
Storage Temperature: Ambient
MDL Number: MFCD00016191
CAS Number: 13760-81-1
EINECS: 237-355-8
UN: 3288
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR