Niobium(V) oxide ≥99.95% (metals basis), sputtering target, Diameter 76.2 mm (3.0 in), Thickness 6.35 mm (0.250 in)

Synonyms: Niobium pentoxide Diniobium pentaoxide

41083-KS 41083-KS
AA41083-KSEA 1001.22 USD
CAAA41083-KS AA41083-KS
Niobium(V) oxide ≥99.95% (metals basis), sputtering target, Diameter 76.2 mm (3.0 in), Thickness 6.35 mm (0.250 in)
Niobium (V) oxide
Formula: Nb₂O₅
MW: 265.81 g/mol
Melting Pt: 1496 °C
Density: 4.6 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011128
CAS Number: 1313-96-8
EINECS: 215-213-6
Merck Index: 14,06559

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR