Chromium(III) oxide ≥99.8% (metals basis), sputtering target, Diameter 76.2 mm (3.0 in), Thickness 3.18 mm (0.125 in)

Warning

Synonyms: Chromic oxide

41058-KS
CAAA41058-KS
Chromium(III) oxide ≥99.8% (metals basis), sputtering target, Diameter 76.2 mm (3.0 in), Thickness 3.18 mm (0.125 in)
Chromium (III) oxide
Formula: Cr₂O₃
MW: 151.99 g/mol
Boiling Pt: 3000 °C
Melting Pt: 2439 °C
Density: 5,22
Storage Temperature: Ambient
MDL Number: MFCD00010949
CAS Number: 1308-38-9
EINECS: 215-160-9
Merck Index: 12,02283

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR