Silicon ≥99.999% (metals basis), sputtering target, Diameter 76.2 mm (3.0 in), Thickness 6.35 mm (0.250 in)

Supplier: Thermo Scientific Chemicals

Warning

40893-KS 40893-KS
AA40893-KSEA 384 USD
CAAA40893-KS AA40893-KS
Silicon ≥99.999% (metals basis), sputtering target, Diameter 76.2 mm (3.0 in), Thickness 6.35 mm (0.250 in)
Silicon
Formula: Si
Boiling Pt: 2355 °C (1013 hPa)
Melting Pt: 1410 °C
Density: 2,34 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00085311
CAS Number: 7440-21-3
EINECS: 231-130-8
Merck Index: 12,08635

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR