Aluminium oxide, aerosol refractory paint

Danger

Synonyms: Alumina C

40391-0CL 40391-CLC
AA40391-0CLEA 883.07 USD
AA40391-0CL AA40391-CLC
Aluminium oxide, aerosol refractory paint
Aluminium oxide

Provides easy application of uniform, thin, protective high temperature Aluminum oxide coatings. Substrates can be ceramic, graphite or metal. Aluminum oxide is also widely used as an abrasive. In the paper-making process.

Formula: Al₂O₃
MW: 101.96 g/mol
Boiling Pt: –44 °C
Melting Pt: 2045 °C
Density: 3.94 g/cm³ (20 °C)
Flash Pt: –104 °C (–155 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003424
CAS Number: 1344-28-1
EINECS: 215-691-6
UN: 1950
ADR: 2.1,
REACH: 01-2119529248-35
Merck Index: 13,00355

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR