Talc
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Magnesium silicate monohydrate
40318-A4
040318-22
40318-36
40318-22
040318-A4
040318-36
AA40318-A4EA
201.9
USD
CAAA40318-A4
CAAA40318-36
AA40318-A4
AA40318-36
CAAA40318-22
AA40318-22
Talc
Talc
Lubricant, filler, electric and heat insulator, in clarifying liquids by filtration
Formula:
Mg₃O₁₁Si₄·1H₂O MW: 379.27 g/mol Melting Pt: >1200 °C Density: 2,7 Storage Temperature: Ambient |
MDL Number:
MFCD00792903 CAS Number: 14807-96-6 EINECS: 238-877-9 Merck Index: 13,09127 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...