Strontium titanate substrate 10x10x1 mm, polished one side 111 orientation

Synonyms: Strontium titanate Strontium titanate (IV) Strontium titanium oxide

40279-KT 40279-KU 40279-KT 40279-KU
AA40279-KTEA 160.95 USD
CAAA40279-KU AA40279-KT CAAA40279-KT AA40279-KU
Strontium titanate substrate 10x10x1 mm, polished one side 111 orientation
Strontium titanate
Formula: O₃SrTi
Melting Pt: 2080 °C
Density: 5.11 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00049554
CAS Number: 12060-59-2
EINECS: 235-044-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR