Aluminium oxide ≥99.9%, single crystal

Synonyms: Alumina C

40265-KJ 40265-KJ
AA40265-KJEA 376.64 USD
CAAA40265-KJ AA40265-KJ
Aluminium oxide ≥99.9%, single crystal
Aluminium oxide
Formula: Al₂O₃
MW: 101.96 g/mol
Boiling Pt: 2980 °C (1013 hPa)
Melting Pt: 2045 °C
Density: 3.94 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003424
CAS Number: 1344-28-1
EINECS: 215-691-6
REACH: 01-2119529248-35
Merck Index: 13,00355

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR