Zirconium(IV) oxide ≥99.5% (metals basis excluding Hf) (max. 100 ppm Hf)

Warning

Synonyms: Zirconium dioxide

40140-36 40140-22 40140-22 40140-36
AA40140-36EA 326.32 USD
AA40140-36 AA40140-22 CAAA40140-36 CAAA40140-22
Zirconium(IV) oxide ≥99.5% (metals basis excluding Hf) (max. 100 ppm Hf)
Zirconium (IV) oxide
Formula: O₂Zr
MW: 123.22 g/mol
Boiling Pt: ∼5000 °C
Melting Pt: ∼2700 °C
Density: 5.89
Storage Temperature: Ambient
MDL Number: MFCD00011310
CAS Number: 1314-23-4
EINECS: 215-227-2
Merck Index: 14,10180

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR