Boron ≥98% (metals basis), crystalline powder -325 mesh

Supplier: Thermo Scientific Chemicals
Warning

040036-09 040036-03 040036-18
AA40036-09EA 158.99 USD
AA40036-09 AA40036-18 AA40036-03
Boron ≥98% (metals basis), crystalline powder -325 mesh
Boron
Formula: B
MW: 10.81 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00134034
CAS Number: 7440-42-8
EINECS: 231-151-2
UN: 3178
ADR: 4.1,III
Merck Index: 13,01333

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR