Trichloroethylene 99+% semiconductor grade

Danger

Synonyms: Trichloroethene 1,1,2-Trichloroethene 1,1,2-Trichloroethylene

39744-K2 39744-K7 39744-K2
AA39744-K2EA 77.86 USD
CAAA39744-K7 CAAA39744-K2 AA39744-K2
Trichloroethylene 99+% semiconductor grade
Trichloroethylene
Formula: ClCH=CCl₂
MW: 131.39 g/mol
Boiling Pt: 87 °C
Melting Pt: –85 °C
Density: 1.46
Storage Temperature: Ambient
MDL Number: MFCD00000838
CAS Number: 79-01-6
EINECS: 201-167-4
UN: 1710
ADR: 6.1,III
Merck Index: 12,09769
https://us.vwr.com/stibo/web/std.lang.all/13/81/741381.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR